Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva rmp Protein A Sepharose™ Fast Flow Affinity Media

Designed for high-purity separation of monoclonal and polyclonal antibodies

Specifications

Binding Capacity >Min. 22 mg Human IgG/mL
Particle Size 60 - 165 µm
Product Type rmp Protein A Sepharose fast flow media
For Use With (Application) For High-Purity Separation of Monoclonal and Polyclonal Antibodies
Ligand Type Recombinant Protein A (E. coli)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
45-002-974
View Documents
Cytiva
17513801
5 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
45-002-975
View Documents
Cytiva
17513802
25 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
45-002-976
View Documents
Cytiva
17513803
200 mL This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Non-mammalian-based media composed of recombinant Protein A coupled with Sepharose 4 Fast Flow base matrix by reductive amination
  • Coupling chemistry that minimizes leakage of protein A ligand
  • Multi-point attachment of rProtein A by chemically-stable amide bonds
  • High capacity and good pressure/flow characteristics provide for high throughput

  • Matrix: 4% cross-linked agarose
  • Average particle size: 90μm (d50V)
  • Ligand: recombinant protein A (E. coli)
  • pH Stability working range: 3 to 10
  • pH Stability cleaning-in-place: 2 to 11
  • Binding capacity/mL chromatography medium: >35mg human IgG/mL
  • Pressure/Flow specification: base matrix 150 to 250cm/hr., 100kPa, XK 50/60 column

For therapeutic applications requiring extremely pure eluate fractions of antibodies with non-exposure of antibodies to human or animal derivatives during purification process.

Specifications

Specifications

>Min. 22 mg Human IgG/mL
rmp Protein A Sepharose fast flow media
Recombinant Protein A (E. coli)
3 to 10
2°C to 8°C
60 - 165 µm
For High-Purity Separation of Monoclonal and Polyclonal Antibodies
4% Cross-Linked Agarose
Stable in aqueous Buffers commonly used in Protein A Chromatography
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit