Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RN 1747, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective TRPV4 agonist (EC50 values are 0.77, 4.0 and 4.1 μM for hTRPV4, mTRPV4 and rTRPV4 respectively). Displays selectivity over other TRP channels (EC50 values are 0.77, >30, >30 and >100 μM for TRPV4, TRPM8, TRPV3 and TRPV1 receptors respectively). Also antagonizes TRPM8 at relevant concentrations (IC50 = 4 μM).
Specifications
Quantity | 50 mg |
Formula Weight | 395.87 |
Percent Purity | >98% |
Chemical Name or Material | RN 1747 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction