Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNA Polymerase II/POLR2A Rabbit anti-Human, Mouse, Clone: 10C7U2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $516.00
Specifications
Antigen | RNA Polymerase II/POLR2A |
---|---|
Clone | 10C7U2 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
RNA Polymerase II/POLR2A Monoclonal antibody specifically detects RNA Polymerase II/POLR2A in Human, Mouse samples. It is validated for Western BlotSpecifications
RNA Polymerase II/POLR2A | |
Western Blot | |
Unconjugated | |
Rabbit | |
Chromatin Research, Epigenetics, Phospho Specific, Transcription Factors and Regulators | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5430 | |
IgG | |
Affinity purified |
10C7U2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2 | |
A phospho specific peptide corresponding to residues surrounding S5 of human RNA Polymerase II/POLR2A | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title