Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ RNA sample loading buffer

Manufacturer:  Alfa Aesar™ J62832XF

 View more versions of this product

Catalog No. AAJ62832XF


Order in Bulk

Request bulk or custom formats

Add to cart

Specifications

Specifications

RNA sample loading buffer
MFCD00213041
Liquid
1mL
Safety and Handling

Safety and Handling

GHS H Statement
H360-H351-H302-H332-H317
May damage fertility or the unborn child.
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Recommended Storage : −30° to −10°C

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit