Learn More
Thermo Scientific™ RNA sample loading buffer, no ethidium bromide
Manufacturer:  Thermo Scientific™ J61937XF
 View more versions of this product
Catalog No. AAJ61937XF
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Formulated for electrophoresis of RNA on formaldehyde-agarose gels with or without ethidium bromide.
Specifications
Specifications
Liquid
1mL
Ambient temperatures