Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA sodium salt, from yeast, Affymetrix/USB™

Click to view available options
Quantity:
25g
100g
1kg
5kg
Packaging:
Plastic bottle
73049-77-1
73049-77-1
Physical Form Powder
Packaging Plastic bottle
Quantity 5kg
Chemical Name or Material RNA sodium salt