Learn More
Andwin Scientific RNASE AWAY
SDP
Manufacturer:  Andwin Scientific E30703
Encompass_Preferred
The RNase AWAY� removes RNase from laboratory surfaces, apparatus, benchtops, glassware, and plastic ware. This product does not leave any residue that may affect gel polymerization or staining and does not consist of any strong acid. It is packaged and sold in a 4 liter refill bottle.
Catalog No. NC9643810
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »