Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNase L Antibody (2E9) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 12 publications

$206.50 - $499.50

Specifications

Antigen RNase L
Clone 2000000000
Concentration 1.0 mg/ml
Dilution Western Blot 1:1000-1:4000 (ECL), ELISA 1:100-1:2000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Knockout Validated
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100351SS
SDP
View Documents
Novus Biologicals
NB100351SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB100351
SDP
View Documents
Novus Biologicals
NB100351
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

RNase L Monoclonal specifically detects RNase L in Human, Hamster, Primate, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

RNase L
1.0 mg/ml
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
Unconjugated
Mouse
Chromatin Research, Cytokine Research, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, Prostate Cancer, Protein Kinase
Q05823
6041
Purified full-length recombinant human RNaseL produced in insect cells. [UniProt# Q05823]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
83 kDa
2000000000
Western Blot 1:1000-1:4000 (ECL), ELISA 1:100-1:2000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Knockout Validated
Monoclonal
Purified
RUO
Human, Hamster, Primate, Mouse (Negative), Rat (Negative)
2-5A-dependent ribonuclease, 2-5A-dependent RNase, 2'-5'-oligoisoadenylate synthetase-dependent, EC 3.1.26.-, HPC1, interferon-induced 2-5A-dependent RNase, MGC104972, MGC133329, PRCA1, prostate cancer 1, Ribonuclease 4, Ribonuclease L, ribonuclease L (2'-5'-oligoisoadenylate synthetase-dependent), RNase L, RNS4DKFZp781D08126
RNASEL
IgG1 κ
Protein G purified
Human, hamster and primate. This antibody does not react with mouse or rat.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.