Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNase11 Monoclonal Antibody (OTI2C11)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527315

Catalog No. PIMA527315


Only null left
Add to Cart

Description

Description

RNase11 Monoclonal Antibody for Western Blot, IHC (P)

RNASE11 (Ribonuclease A Family Member 11 (Inactive)) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and ribonuclease activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RNase11
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8TAA1
RNASE11
Full length human recombinant protein of RNASE11 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI2C11
Unconjugated
RNASE11
C14orf6; epididymis secretory sperm binding protein Li 84p; HEL-S-84p; Probable ribonuclease 11; putative inactive ribonuclease 11; RAJ1; ribonuclease 11; ribonuclease A family member 11 (inactive); ribonuclease A j1; ribonuclease, RNase A family, 11 (non-active); RNase 11; RNASE11; UNQ5832/PRO19669
Mouse
Affinity Chromatography
RUO
122651
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.