Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNASE13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575903
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Does not exhibit any ribonuclease activity. [UniProt].
Specifications
RNASE13 | |
Polyclonal | |
Unconjugated | |
RNASE13 | |
epididymis secretory sperm binding protein Li 86p; HEL-S-86p; pancreatic ribonuclease; probable inactive ribonuclease-like protein 13; RAL1; ribonuclease 13; ribonuclease A family member 13 (inactive); ribonuclease A L1; ribonuclease, RNase A family, 13 (non-active); ribonuclease-like protein 13; Rnase13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
440163, 497071, 497194 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5GAL7, Q5GAM7, Q5GAN3 | |
RNASE13 | |
Recombinant full length Human RNASE13. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction