Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNASE13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110302
Description
Immunogen sequence: MAPAVTRLLF LQLVLGPTLV MDIKMQIGSR NFYTLSIDYP RVNYPKGFRG YCNGLMSYMR GKMQNSDCPK IHYVIHAPWK AIQKFCKYSD SFCENYNEYC TLTQDSLPIT VCSLSHQQPP TSCYYNSTLT NQKLYLLCSR KYEADPIGIA GLYSGI.
Does not exhibit any ribonuclease activity. [UniProt].
Specifications
RNASE13 | |
Polyclonal | |
Unconjugated | |
RNASE13 | |
epididymis secretory sperm binding protein Li 86p; HEL-S-86p; pancreatic ribonuclease; probable inactive ribonuclease-like protein 13; RAL1; ribonuclease 13; ribonuclease A family member 13 (inactive); ribonuclease A L1; ribonuclease, RNase A family, 13 (non-active); ribonuclease-like protein 13; Rnase13 | |
Rabbit | |
Affinity chromatography | |
RUO | |
440163, 497071 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5GAM7, Q5GAN3 | |
RNASE13 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-156 of human RNASE13 (NP_001012264.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction