Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNASE3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548595

Catalog No. PA548595


Add to cart

Description

Description

Cytotoxin and helminthotoxin with low-efficiency ribonuclease activity. Possesses a wide variety of biological activities. Exhibits antibacterial activity, including cytoplasmic membrane depolarization of preferentially Gram-negative, but also Gram-positive strains. Promotes E.coli outer membrane detachment, alteration of the overall cell shape and partial loss of cell content.
Specifications

Specifications

RNASE3
Polyclonal
PBS with 0.09% sodium azide
P12724
Eosinophil cationic protein, ECP, 3127-, Ribonuclease 3, RNase 3, RNASE3, ECP, RNS3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6037
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
RNASE3
Liquid
RNASE3
KLH conjugated synthetic peptide between 124-151 amino acids from the C-terminal region of human RNASE3.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit