Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNASEH2B Mouse anti-Human, Biotin, Clone: 673, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243776B

Catalog No. NBP243776B


Add to cart

Description

Description

RNASEH2B Monoclonal antibody specifically detects RNASEH2B in Human samples. It is validated for Western Blot.
Specifications

Specifications

RNASEH2B
673
Western Blot
Purified
RNASEH2B
Recombinant protein encompassing a sequence within the center region of human RNaseH2B. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot
Biotin
PBS with 0.05% Sodium Azide
ribonuclease H2, subunit B
Mouse
IgG1
0.1 ml
Primary
79621
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit