Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNaseH2B Monoclonal Antibody (GT673), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527850
Description
Positive Control: HeLa, HeLa nuclear Predicted Reactivity: Pig (85%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RNaseH2B Monoclonal antibody specifically detects RNaseH2B in Human samples. It is validated for Western Blot.Specifications
RNaseH2B | |
Monoclonal | |
2.01 mg/mL | |
PBS with no preservative | |
Q5TBB1 | |
RNASEH2B | |
Recombinant protein encompassing a sequence within the center region of human RNaseH2B. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
GT673 | |
Unconjugated | |
RNASEH2B | |
1110019N06Rik; 2610207P08Rik; AGS2; Aicardi-Goutieres syndrome 2 protein; Deleted in lymphocytic leukemia 8; deleted in lymphocytic leukemia 8 homolog; DLEU8; ribonuclease H2; ribonuclease H2 subunit B; ribonuclease H2, subunit B; ribonuclease HI subunit B; RNase H2 subunit B; Rnaseh2b | |
Mouse | |
Protein G | |
RUO | |
79621 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction