Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNaseH2B Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA580974
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human RNASEH2B.
RNASEH2B is composed of a single catalytic subunit (A) and two non-catalytic subunits (B and C) and specifically degrades the RNA of RNA:DNA hybrids. RNASEH2B is the non-catalytic B subunit of RNase H2, which is thought to play a role in DNA replication.
Specifications
RNaseH2B | |
Polyclonal | |
PBS with no preservative | |
Q5TBB1 | |
RNASEH2B | |
E. coli-derived Human RNASEH2B fragment | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RNASEH2B | |
AGS2, DLEU8, Aicardi-Goutieres syndrome 2 protein, RNase H2 subunit B, deleted in lymphocytic leukemia 8, ribonuclease H2 subunit B, ribonuclease HI subunit B | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
79621 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction