Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RND1 Mouse anti-Human, Clone: OTI1C3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525234

Catalog No. PIMA525234


Add to cart

Description

Description

Lacks intrinsic GTPase activity. Has a low affinity for GDP, and constitutively binds GTP. Controls rearrangements of the actin cytoskeleton. Induces the Rac-dependent neuritic process formation in part by disruption of the cortical actin filaments. Causes the formation of many neuritic processes from the cell body with disruption of the cortical actin filaments.
Specifications

Specifications

RND1
Monoclonal
0.36 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q92730
RHO6
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27289
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1C3
Unconjugated
RND1
Liquid
RND1
Full length human recombit protein of RND1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit