Learn More
RNF112 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 251651AP
Catalog No. 50-173-4762
Add to cart
Description
Description
The specific function of this protein remains unknown. This gene encodes a member of the RING finger protein family of transcription factors. The protein is primarily expressed in brain. The gene is located within the Smith-Magenis syndrome region on chromosome 17.
Specifications
Specifications
RNF112
0.49 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96DY5, Q9ULX5
BFP, ring finger protein 112, RNF112, ZNF179
Rabbit
IgG
150 μL
-20°C
Primary
22671, 7732
Western Blot
Unconjugated
RNF112
Liquid
RNF112
RNF112 Fusion Protein Ag18306
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse