Learn More
RNF121 Mouse anti-Human, Clone: 2G7, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524471
Catalog No. PIMA524471
Add to cart
Description
Description
Peptide Sequence: ERDFAEMCAD YMASTIGFYS ESGMPTKHLS DSVCAVCGQQ IFVDVSEEGI IENTYRLSCN HVFHEFCIRG WCIVGKKQTC PYCKEKVDLK RMFSNPWERP HVMYGQLLD
The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Several alternatively spliced transcript variants have been noted for this gene, however, not all are likely to encode viable protein products.
Specifications
Specifications
RNF121
Monoclonal
Unconjugated
RNF121
Liquid
RNF121
RNF121 (NP_060790.2, 193 a.a. ∽ 301 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
2G7
PBS with no preservative; pH 7.4
Q9H920
RNF121
Mouse
IgG1, kappa
100 μg
-20° C
Primary
55298