Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF121 Polyclonal Antibody
RNF121 Polyclonal Antibody
Supplier: Thermo Scientific PA572876
Description
RNF121 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Specifications
RNF121 | |
Polyclonal | |
Unconjugated | |
RNF121 | |
4930544L10Rik; RING finger protein 121; Rnf121 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
308871, 55298, 75212 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8R1Z9, Q9H920 | |
RNF121 | |
RNF121 antibody for a 19 amino acid peptide near the carboxy terminus of human RNF121. The immunogen is within amino acids 260 - 310 of RNF121. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction