Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF122 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549075

Catalog No. PA549075

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

May induce necrosis and apoptosis. May play a role in cell viability.
Specifications

Specifications

RNF122
Polyclonal
PBS with 0.09% sodium azide
Q9H9V4
RING finger protein 122, RNF122
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79845
Western Blot
Unconjugated
RNF122
Liquid
RNF122
KLH conjugated synthetic peptide between 84-110 amino acids from the Central region of human RNF122.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit