Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF138 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101572
Description
Antibody detects endogenous levels of total RNF138.
The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-DNA and protein-protein interactions. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq, Jul 2008].
Specifications
RNF138 | |
Polyclonal | |
Unconjugated | |
Rnf138 | |
2410015A17Rik; 2810480D20Rik; E3 ubiquitin-protein ligase RNF138; hNARF; HSD4; HSD-4; NARF; nemo-like kinase associated ring finger protein; Nemo-like kinase-associated RING finger protein; NLK-associated RING finger protein; ring finger protein 138; ring finger protein 138, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF138; Rnf138; Rsd4; RSD-4; STRIN; testicular tissue protein Li 165; Trif; Trif-d | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51444, 56515, 94196 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8WVD3, Q99PD2, Q9CQE0 | |
Rnf138 | |
A synthesized peptide derived from human RNF138(Accession Q8WVD3), corresponding to amino acid residues N132-F182. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction