Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF139 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159756
Description
RNF139 Polyclonal specifically detects RNF139 in Human samples. It is validated for Western Blot.Specifications
RNF139 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q8WU17 | |
RNF139 | |
Synthetic peptides corresponding to RNF139(ring finger protein 139) The peptide sequence was selected from the N terminal of RNF139. Peptide sequence SQRSLFKFYTYSSAFLLAATSVLVNYYASLHIDFYGAYNTSAFGIELLPR. | |
100 μL | |
Zinc Finger | |
11236 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS & 2% Sucrose. with 0.09% Sodium Azide | |
EC 6.3.2.-, HRCA1, patched related protein translocated in renal cancer, RCA1multiple membrane spanning receptor TRC8, ring finger protein 139MGC31961, Translocation in renal carcinoma on chromosome 8 protein, TRC8E3 ubiquitin-protein ligase RNF139 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Pig: 100%; Rat: 100%; Equine: 92%; Rabbit: 86%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title