Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568613
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein interacts with androgen receptor (AR) and may function as a coactivator that induces AR target gene expression in prostate. A dominant negative mutant of this gene has been demonstrated to inhibit the AR-mediated growth of prostate cancer. This protein also interacts with class III ubiquitin-conjugating enzymes (E2s) and may act as a ubiquitin-ligase (E3) in the ubiquitination of certain nuclear proteins. Five alternatively spliced transcript variants encoding two distinct isoforms have been reported.
Specifications
RNF14 | |
Polyclonal | |
Unconjugated | |
RNF14 | |
2310075C09Rik; 2610005D23Rik; AA986456; androgen receptor associated protein 54; androgen receptor associated protein 54 (androgen receptor coactivator); androgen receptor-associated protein 54; ARA54; AU041447; D18Ertd188e; D7Bwg0165e; E3 ubiquitin-protein ligase RNF14; HFB30; HRIHFB2038; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase RNF14; protein Triad2; RING finger protein 14; RNF14; TRIAD2; Triad2 protein; TRIAD2 type I | |
Rabbit | |
Affinity chromatography | |
RUO | |
619577 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
RNF14 | |
synthetic peptide corresponding to a region of rat RNF14. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction