Learn More
TRIM38 Mouse anti-Canine, Human, Mouse, Non-human primate, Rat, Clone: OTI15A3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA526235
Catalog No. PIMA526235
Add to cart
Description
Description
This gene encodes a member of the tripartite motif (TRIM) family. The encoded protein contains a RING-type zinc finger, B box-type zinc finger and SPRY domain. The function of this protein has not been identified. A pseudogene of this gene is located on the long arm of chromosome 4.
Specifications
Specifications
TRIM38
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O00635
E3 ubiquitin-protein ligase TRIM38; 632-; RING finger protein 15; Tripartite motif-containing protein 38; Zinc finger protein RoRet; TRIM38; RNF15; RORET
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10475, 214158, 488257, 680019, 694861
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI15A3
Unconjugated
TRIM38
Liquid
TRIM38
Human recombit protein fragment corresponding to amino acids 1-265 of TRIM38 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Non-human primate, Rat