Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF160 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Specifications
Antigen | RNF160 |
---|---|
Concentration | 367 μg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8702
|
Proteintech Group Inc
284521AP150UL |
150 μL |
Each of 1 for $496.07
|
N/A | |||||
Description
The RNF160 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RNF160. This antibody recognizes human, rat antigen. The RNF160 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
RNF160 | |
Western Blot | |
Unconjugated | |
Human, Rat | |
IgG |
367 μg/mL | |
Polyclonal | |
Rabbit | |
26046 | |
Primary |
Spot an opportunity for improvement?
Product Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title