Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF165 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548894

Catalog No. PA548894

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Encoded in regions involved in pericentric inversions in patients with bipolar affective disorder. [provided by RefSeq, Jul 2008]
Specifications

Specifications

RNF165
Polyclonal
Unconjugated
RNF165
Liquid
RNF165
KLH conjugated synthetic peptide between 3-31 amino acids from the N-terminal region of human RNF165.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q6ZSG1
RING finger protein 165, RNF165
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
494470
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit