Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ RNF168, Rabbit, Unlabeled, Polyclonal,
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ 061130I
Description
E3 ubiquitin-protein ligase RNF168 (UniProt Q8IYW5; also known as hRNF168, RING finger protein 168) is encoded by the RNF168 gene (Gene ID 165918) in human. Ubiquitylation/Ubiquitination is an essential early signal in the DNA damage repair process. Multiple E3 ligases, including RNF2 (RING2), RNF8 and RNF168, mediate the addition of K63-linked polyubiquitin chains to histone 2A (H2A) and H2AX, which in turn recruits DNA damage repair proteins, e.g. RAP80, BRCA1. In addition, SUMOylation and NEDDylation are also involved in the DNA damage response. RNF168 is an a dual specificity E3 ligase that mediates both the ubiquitination and NEDDylation of H2A and H2AX. The two types of modifications compete against each other and NEDD8 modification of H2A and H2AX blocks the recruitment of BRCA1 at DNA damage repair sites. RNF168 itself is also subjected to NEDD8 modification and NEDDylation of RNF168 is necessary for its ubiquitin ligase activity. Inhibition of RNF168 NEDDylation impairs its interaction with E2 enzyme Ubc13 (UBE2N). Likewise, downregulating RNF168 NEDDylation by the deNEDDylating enzyme NEDP1, or due to mutations of the NEDD8-conjugating enzyme UBC12, decreases H2A and H2AX ubiquitylation. RNF168 Mutations are linked to RIDDLE (radiosensitivity, immunodeficiency dysmorphic features and learning difficulties) syndrome.Specifications
RNF168 | |
Polyclonal | |
Unconjugated | |
RNF168 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This polyclonal antibody detected nuclear RNF168 immunoreactivity in 4% paraformaldehyde-fixed, 0.3% Triton X-100-permeabilized human B-cell hybridoma SA13. | |
Stable for 1 year at 2°-8°C from date of receipt. |
Immunoprecipitation, Western Blot | |
Please refer to lot specific datasheet. | |
Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide. | |
Q8IYW5 | |
GST-tagged full-length human RNF168. | |
100 μL | |
Epigenetics & Nuclear Function | |
NP_689830 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction