Learn More
RNF17 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 243021AP
Catalog No. 50-173-4289
Add to cart
Description
Description
RNF17 also termed as ring finger protein 17 is a 1623 amino acid protein, which contains 1 ring type zinc finger and 4 tudor domains. RNF17 is specifically expressed in testis. The RING finger motif presents in many ubiquitin E3 ligases. RNF17 interacts with all four members of the Mad family (Mad1, Mxi1, Mad3 and Mad4), which are basic-helix-loop-helix-leucine zipper transcription factors of the Myc oncoprotein network. RNF17 is component of the tectonic-like complex, which is required for ciliogenesis and sonic hedgehog/SHH signaling. RNF17 is able to form dimers or polymers both in vitro and in vivo, indicating that it may play a role in the assembly of RNF17 granules. RNF17 encodes a novel key regulator of spermiogenesis.
Specifications
Specifications
RNF17
0.62 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q99MV7, Q9BXT8
Mmip 2, ring finger protein 17, RNF17, SPATA23, TDRD4
Rabbit
IgG
150 μL
-20°C
Primary
30054, 56163
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RNF17
Liquid
Rnf17
RNF17 Fusion Protein Ag20144
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse