Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF182 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549229

Catalog No. PA549229

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

E3 ubiquitin-protein ligase that mediates the ubiquitination of ATP6V0C and targets it to degradation via the ubiquitin-proteasome pathway.
Specifications

Specifications

RNF182
Polyclonal
PBS with 0.09% sodium azide
Q8N6D2
E3 ubiquitin-protein ligase RNF182, 632-, RING finger protein 182, RNF182
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
221687
Western Blot
Unconjugated
RNF182
Liquid
RNF182
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human RNF182.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit