Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF186 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | RNF186 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15977420
|
Novus Biologicals
NBP15977420UL |
20 μL |
Each for $204.00
|
|
|||||
NBP159774
|
Novus Biologicals
NBP159774 |
100 μL |
Each for $482.50
|
|
|||||
Description
RNF186 Polyclonal specifically detects RNF186 in Human samples. It is validated for Western Blot.Specifications
RNF186 | |
Polyclonal | |
Rabbit | |
Q9NXI6 | |
54546 | |
Synthetic peptides corresponding to RNF186(ring finger protein 186) The peptide sequence was selected from the middle region of RNF186. Peptide sequence GQLAQPCTEVSLCPQGLVDPADLAAGHPSLVGEDGQDEVSANHVAARRLA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
FLJ20225, ring finger protein 186, RP11-91K11.1 | |
RNF186 | |
IgG |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title