Learn More
RNF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 160311AP
Catalog No. 50-173-0660
Add to cart
Description
Description
The protein encoded by this gene is a molecular chaperone that is a member of the chaperonin containing TCP1 complex , also known as the TCP1 ring complex. This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of this gene, encoding different isoforms, have been characterized. In addition, three pseudogenes that appear to be derived from this gene have been found.
Specifications
Specifications
RNF2
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4KLY4, Q99496, Q9CQJ4
BAP 1, BAP1, DING, HIP2 interacting protein 3, HIPI3, Protein DinG, RING finger protein 1B, ring finger protein 2, RING finger protein BAP 1, RING1B, RING2, RNF2
Rabbit
IgG
150 μL
-20°C
Primary
19821, 304850, 6045
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
RNF2
Liquid
RNF2
RNF2 Fusion Protein Ag8900
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat