Learn More
RNF212 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 211781AP
Catalog No. 50-173-3301
Add to cart
Description
Description
This gene encodes a RING finger protein that may function as a ubiquitin ligase. The encoded protein may be involved in meiotic recombination. This gene is located within a linkage disequilibrium block and polymorphisms in this gene may influence recombination rates. Alternate splicing results in multiple transcript variants.
Specifications
Specifications
RNF212
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
F6TQD1, Q495C1
ring finger protein 212, RNF212, ZHP3
Rabbit
IgG
150 μL
-20°C
Primary
285498, 671564, 689721
Western Blot
Unconjugated
RNF212
Liquid
RNF212
RNF212 Fusion Protein Ag15457
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat