Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF213 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551902
Description
Immunogen sequence: VGKNEQGEPE DLKKPEGKNR SAAAVKNEKE QKNQEADVQE VKASTLSPGG GVTVFFHAII SLHFPFNPDL HKVFIRGGEE FGESKWDSNI CELHYTRDLG HDRVLVEGIV CISKKHLDKY IPYKYVIYNG ESFEYEFIYK H Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 48%.
RNF213 is an E3 ubiquitin-protein ligase involved in angiogenesis. Involved in the non-canonical Wnt signaling pathway in vascular development: acts by mediating ubiquitination and degradation of FLNA and NFATC2 downstream of RSPO3, leading to inhibit the non-canonical Wnt signaling pathway and promoting vessel regression. Also has ATPase activity.
Specifications
RNF213 | |
Polyclonal | |
Unconjugated | |
RNF213 | |
6030403J01; ALK lymphoma oligomerization partner on chromosome 17; ALO17; C17orf27; D11Ertd759e; E3 ubiquitin-protein ligase RNF213; KIAA1554; KIAA1618; mKIAA1554; MYMY2; Mysterin; MYSTR; NET57; protein ALO17; RGD1308168; RING finger protein 213; RING-type E3 ubiquitin transferase RNF213; RNF213 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57674 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q63HN8 | |
RNF213 | |
Recombinant protein corresponding to Human RNF213. Recombinant protein control fragment (Product #RP-102049). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction