Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF213 Rabbit anti-Human, Polyclonal , Abnova™

Rabbit polyclonal antibody raised against recombinant human RNF213.

Manufacturer:  Abnova Corporation PAB29398

Catalog No. 89-934-862


Add to cart

Description

Description

Sequence: VGKNEQGEPEDLKKPEGKNRSAAAVKNEKEQKNQEADVQEVKASTLSPGGGVTVFFHAIISLHFPFNPDLHKVFIRGGEEFGESKWDSNICELHYTRDLGHDRVLVEGIVCISKKHLDKYIPYKYVIYNGESFEYEFIYKH
Specifications

Specifications

RNF213
Unconjugated
Immunohistochemistry (1:200-1:500) Immunofluorescence (1-4 ug/mL) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user.
Liquid
RNF213
Recombinant protein corresponding to human RNF213.
Antigen affinity purification
RUO
Primary
57674
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against recombinant human RNF213.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
C17orf27/KIAA1554
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit