Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF39 Mouse anti-Human, Mouse, Clone: OTI5F5, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA505915
Description
This gene lies within the major histocompatibility complex class I region on chromosome 6. Studies of a similar rat protein suggest that this gene encodes a protein that plays a role in an early phase of synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
RNF39 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9H2S5 | |
HZF, HZFW, LIRF | |
Mouse | |
IgG2b | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
386454, 80352 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI5F5 | |
Unconjugated | |
RNF39 | |
Liquid | |
RNF39 | |
Full length human recombit protein of human RNF39 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human, Mouse |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title