Learn More
RNF39 Mouse anti-Canine, Human, Mouse, Non-human primate, Rat, Clone: OTI5E10, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA526227
Catalog No. PIMA526227
Add to cart
Description
Description
This gene lies within the major histocompatibility complex class I region on chromosome 6. Studies of a similar rat protein suggest that this gene encodes a protein that plays a role in an early phase of synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
RNF39
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H2S5, Q920M2
RNF39; LIRF; ring finger protein 39; HZFW1; HZFw1 protein; LTP (long-term potentiation) induced RING finger protein; DADB-60O11.8; HZF; HZFW
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
171387, 386454, 488344, 712215, 80352
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI5E10
Unconjugated
RNF39
Liquid
RNF39
Full length human recombit protein of RNF39 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Non-human primate, Rat