Learn More
RNF40 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 156211AP
Catalog No. 50-173-0357
Add to cart
Description
Description
The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein was reported to interact with the tumor suppressor protein RB1. Studies of the rat counterpart suggested that this protein may function as an E3 ubiquitin-protein ligase, and facilitate the ubiquitination and degradation of syntaxin 1, which is an essential component of the neurotransmitter release machinery. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
RNF40
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75150, Q3U319, Q8CJB9
BRE1 B, BRE1B, KIAA0661, RBP95, ring finger protein 40, RNF40, STARING
Rabbit
IgG
150 μL
-20°C
Primary
233900, 266712, 9810
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
RNF40
Liquid
RNF40
RNF40 Fusion Protein Ag8047
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat