Learn More
RNF43 Mouse anti-Human, Alexa Fluor 488, Clone: 923227R, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB7964G100UG
Catalog No. FAB7964G100
Add to cart
This item is not returnable. View return policy
Description
Description
RNF43 Monoclonal antibody specifically detects RNF43 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
RNF43
923227R
Flow Cytometry 0.25-1 μg/10^6 cells
Q68DV7
DKFZp781H02126, DKFZp781H0392, FLJ20315, FLJ77466, FLJ99338, MGC125630, ring finger protein 43, RNF124, URCC
Human embryonic kidney cell line HEK293-derived recombinant human RNF43 Glu43-Val199 Accession # Q68DV7
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human RNF43 in direct ELISAs.
Flow Cytometry
Alexa Fluor 488
Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG1
100 μg
Primary
54894
Human