Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566741
Description
Immunogen sequence: NAHEPFRRGTG VDLGQGHPAS SW Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
The protein encoded by this gene contains a RING finger, which is a motif known to be involved in protein-protein interactions. This protein is a membrane-bound ubiquitin ligase. It can regulate cell motility by targeting paxillin ubiquitination and altering the distribution and localization of paxillin in cytoplasm and cell focal adhesions.
Specifications
RNF5 | |
Polyclonal | |
Unconjugated | |
RNF5 | |
2410131O05Rik; AA407576; E3 ubiquitin-protein ligase RNF5; G16; HsRma1; ing finger protein 5 pseudogene; NG2; Protein G16; ram1 homolog; RING finger protein 5; ring finger protein 5, E3 ubiquitin protein ligase; RING5; RING-type E3 ubiquitin transferase RNF5; RMA1; RNF5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6048 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99942 | |
RNF5 | |
Recombinant Human RNF5. Recombinant protein control fragment (Product #RP-107397). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction