Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | RNF6 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB8025120UL
![]() |
Novus Biologicals
NBP18025120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180251
![]() |
Novus Biologicals
NBP180251 |
100 μL |
Each for $487.50
|
|
|||||
Description
RNF6 Polyclonal specifically detects RNF6 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RNF6 | |
Polyclonal | |
Purified | |
RUO | |
DKFZp686P0776, E3 ubiquitin-protein ligase RNF6, EC 6.3.2.-, ring finger protein (C3H2C3 type) 6, RING-H2 protein RNF-6, SPG2 | |
RNF6 | |
IgG | |
Protein A purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
NP_083050 | |
6049 | |
Synthetic peptide directed towards the N terminal of mouse RNF6. Peptide sequence MDPSRSRSGGSGEESSFQENERRWQQERLHREEAYYQFINELSDEDYRLM. | |
Primary | |
73 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title