Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen RNF7
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093270
View Documents
Novus Biologicals
NBP304412100UL
100 μg Each for $320.71
Add to cart
 
NB093271
View Documents
Novus Biologicals
NBP30441220UL
20 μg Each for $138.37
Add to cart
 
Description

Description

RNF7 Polyclonal antibody specifically detects RNF7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

RNF7
Unconjugated
Rabbit
Human, Mouse, Rat
CKBBP1zinc RING finger protein SAG, CKII beta-binding protein 1, Rbx2, Regulator of cullins 2, ring finger protein 7rbx2, ROC2RING-box protein 2, SAGsensitive to apoptosis, zinc RING finger protein SAG, regulator of cullins 2, Sensitive to apoptosis gene protein
Recombinant fusion protein containing a sequence corresponding to amino acids 1-113 of human RNF7 (NP_055060.1).
Polyclonal
Affinity purified
Immunofluorescence, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
9616
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit