Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF92 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569231
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the nucleus and the cytoplasm. Its function has not been identified.
Specifications
RNF92 | |
Polyclonal | |
Unconjugated | |
TRIM11 | |
BIA1; E3 ubiquitin-protein ligase TRIM11; Protein BIA1; RING finger protein 92; RING-type E3 ubiquitin transferase TRIM11; RNF92; Trim11; tripartite motif containing 11; tripartite motif protein 11; tripartite motif-containing 11; tripartite motif-containing protein 11 | |
Rabbit | |
Affinity chromatography | |
RUO | |
94091 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q99PQ2 | |
TRIM11 | |
synthetic peptide corresponding to a region of mouse RNF92 (aa 383-432). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction