Learn More
RNFT1 Mouse anti-Human, Clone: 4H7, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524462
Catalog No. PIMA524462
Add to cart
Description
Description
Peptide Sequence: MQANRSQLHS PPGTGSSEDA STPQCVHTRL TGEGSCPHSG DVHIQINSIP KECAENASSR NIRSGVHSCA HGCVHSRLRG HSHSEARLTD DTAAESGDHG
Specifications
Specifications
RNFT1
Monoclonal
Unconjugated
RNFT1
Liquid
RNFT1
LOC51136 (NP_057209, 1 a.a. ∽ 100 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
4H7
PBS with no preservative; pH 7.4
Q5M7Z0
RING finger and transmembrane domain-containing protein 1, Protein PTD016, RNFT1
Mouse
IgG2a, kappa
100 μg
-20° C
Primary
51136