Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ RNGTT Human Recombinant Protein
Highly purified. Generating reliable and reproducible results.
$344.00 - $976.00
Specifications
Concentration | 0.25mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | 20mM Tris-HCl buffer (pH 7.5) containing 0.2M NaCl, 40% glycerol, 2mM DTT, 0.1mM PMSF |
Gene ID (Entrez) | 8732 |
Molecular Weight | TMW: 70.9kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25181705
|
Novus Biologicals™
NBP2518170.05MG |
0.05mg |
Each for $344.00
|
|
NBP25181725
|
Novus Biologicals™
NBP2518170.25MG |
0.25mg |
Each for $976.00
|
|
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-597 of Human RNGTT The Recombinant Human RNGTT Protein is derived from E. coli. The Recombinant Human RNGTT Protein has been validated for the following applications: SDS-Page.Specifications
0.25mg/mL | |
20mM Tris-HCl buffer (pH 7.5) containing 0.2M NaCl, 40% glycerol, 2mM DTT, 0.1mM PMSF | |
TMW: 70.9kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
RNGTT | |
Yes |
SDS-Page | |
8732 | |
>85%, by SDS-PAGE | |
CAP1AmRNA-capping enzyme, hCAP, HCE1DKFZp686J2031, HCEHCAP1, RNA guanylyltransferase and 5'-phosphatase | |
Unlabeled |
For research use only and is not approved for use in humans or in clinical diagnosis
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title