Learn More
Aviva Systems Biology Corp RNH1 Antibody - middle region (ARP57822_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP57822_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNH1. Alternate names: MGC18200, MGC4569, MGC54054, RAI, RNHApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62160