Learn More
Invitrogen™ RNMT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113700
Description
Immunogen sequence: ETEDVPKDKS STGDGTQNKR KIALEDVPEK QKNLEEGHSS TVAAHYNELQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Rabbit: 79%.
RNMT belongs to the mRNA cap methyltransferase family. RNMT is a mRNA-capping methyltransferase that methylates the N7 position of the added guanosine to the 5'-cap structure of mRNAs. It binds RNA containing 5'-terminal GpppC.
Specifications
RNMT | |
Polyclonal | |
Unconjugated | |
Rnmt | |
2610002P10Rik; AI848273; cm1p; CMT1; CMT1c; hcm1p; hCMT1; hMet; Kiaa0398; MET; mKIAA0398; mRNA (guanine-7-)methyltransferase; mRNA (guanine-N(7)-)-methyltransferase; mRNA cap guanine-N7 methyltransferase; mRNA cap methyltransferase; RG7MT1; RNA (guanine-7-) methyltransferase; RNA guanine-7 methyltransferase; RNMT | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8731 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O43148 | |
Rnmt | |
Synthetic peptide directed towards the N terminal region of human RNMT (aa 108-157). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.