Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNPC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565724
Description
Immunogen sequence: MAAPEQPLAIS RGCTSSSSLS PPRGDRTLLV RHLPAELTAE EKEDLLKYFG AQSVRVLSDK GRLKHTA Highest antigen sequence identity to the following orthologs - mouse 87%, rat 85%.
Participates in pre-mRNA U12-dependent splicing, performed by the minor spliceosome which removes U12-type introns. U12-type introns comprises less than 1% of all non-coding sequences. Binds to the 3'-stem-loop of m(7)G-capped U12 snRNA.
Specifications
RNPC3 | |
Polyclonal | |
Unconjugated | |
RNPC3 | |
2810441O16Rik; AI447568; C030014B17Rik; KIAA1839; Rbm40; RNA binding region (RNP1, RRM) containing 3; RNA recognition protein; RNA-binding motif protein 40; RNA-binding protein 40; RNA-binding region (RNP1, RRM) containing 3; RNA-binding region-containing protein 3; RNP; Rnpc3; SNRNP65; U11/U12 small nuclear ribonucleoprotein 65 kDa protein; U11/U12 snRNP 65 kDa protein; U11/U12 snRNP 65K; U11/U12-65K | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55599 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96LT9 | |
RNPC3 | |
Recombinant Human RNPC3. Recombinant protein control fragment (Product #RP-106397). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction