Learn More
Aviva Systems Biology Corp RNPEPL1 Antibody - N-terminal region (ARP57162_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP57162_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNPEPL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNPEPL1. Alternate names: FLJ10806, FLJ26675, MGC99544ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-62097