Learn More
RNPEPL1 Mouse anti-Non-species specific, DyLight 755, Clone: 4A9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259371IR
Catalog No. NBP259371IR
Add to cart
Description
Description
RNPEPL1 Monoclonal antibody specifically detects RNPEPL1 in Non-species specific samples. It is validated for Western Blot.
Specifications
Specifications
RNPEPL1
4A9
Western Blot
Purified
RNPEPL1
Synthetic N-terminal arginylated peptide conjugated to KLH
Protein G purified
Store at 4C in the dark.
Monoclonal
Specific for N-terminal arginine next to glutamic acid. Does not detect N-terminal arginine next to aspartic acid or internal arginine residues.
Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
argininyl aminopeptidase-like 1, arginyl aminopeptidase (aminopeptidase B)-like 1, arginyl aminopeptidase-like 1, EC 3.4.11.-, FLJ10806, FLJ26675, MGC99544, RNPEP-like 1
Mouse
IgG1
100 ul
Primary
57140
Non-species specific