Learn More
RNPEPL1 Mouse anti-Non-species specific, HRP, Clone: 2A4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259370H
Catalog No. NBP259370H
Add to cart
Description
Description
RNPEPL1 Monoclonal antibody specifically detects RNPEPL1 in Non-species specific samples. It is validated for Western Blot.
Specifications
Specifications
RNPEPL1
2A4
Western Blot
Purified
RNPEPL1
Synthetic N-terminal arginylated peptide conjugated to KLH
Protein G purified
Store at 4C in the dark.
Monoclonal
Specific for N-terminal arginine, next to both glutamic acid and aspartic acid. Does not detect internal arginine.
Western Blot
HRP
PBS with No Preservative
argininyl aminopeptidase-like 1, arginyl aminopeptidase (aminopeptidase B)-like 1, arginyl aminopeptidase-like 1, EC 3.4.11.-, FLJ10806, FLJ26675, MGC99544, RNPEP-like 1
Mouse
IgG3
100 ul
Primary
57140
Non-species specific